Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024786.1 | NP_001019957.1 | 3 | 412 | ||
XM_008763872.1 | XP_008762094.1 | 3 | 407 | |||
XM_008763873.1 | XP_008762095.1 | 3 | 407 | |||
GenBank mRNA | BC083631.1 | - | 3 | 392 | ||
BC097402.1 | - | 3 | 412 | 7453940 |