Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009424.1 | NP_001009424.1 | 11 | 1004 | ||
GenBank mRNA | AB262963.1 | - | 11 | 860 | ||
AY190609.1 | - | 11 | 1004 | |||
FQ223380.1 | - | 992 |