Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134873.1 | NP_001128345.1 | 7 | 818 | ||
XM_006238781.2 | XP_006238843.1 | 7 | 811 | |||
GenBank mRNA | BC161954.1 | - | 7 | 796 | 9027103 | |
FQ210258.1 | - | 6 | 726 | |||
FQ217494.1 | - | 7 | 776 |