Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmpste24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107974.1 | NP_001101444.1 | 10 | 1606 | ||
GenBank mRNA | FQ211274.1 | - | 1347 |