Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp11l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107978.2 | NP_001101448.1 | 2, 3 | 197 | ||
GenBank mRNA | BC168964.1 | - | 2, 3 | 197 | 7130611 | |
FQ226285.1 | - | 2, 3 | 209 | |||
FQ231750.1 | - | 2, 3 | 218 | |||
FQ233930.1 | - | 2, 3 | 209 |