Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199108.1 | NP_954539.1 | 2 | 301 | ||
GenBank mRNA | BC061837.1 | - | 2 | 301 | 5599940 |