Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037199.3 | NP_001032276.3 | 1 | 349 | ||
GenBank mRNA | BC107450.1 | - | 1 | 349 | 7322355 | |
EF125189.1 | - | 1 | 795 |