Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_233702.7 | XP_233702.4 | 10 | 1795 | ||
GenBank mRNA | BC098821.1 | - | 4 | 528 |