Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277226.1 | NP_001264155.1 | 17 | 2806 | ||
NM_001277227.1 | NP_001264156.1 | 17 | 2662 | |||
NM_003667.3 | NP_003658.1 | 18 | 2878 | |||
NR_110596.1 | 20 | 2580 | ||||
GenBank mRNA | AB105190.1 | - | 18 | 2847 | ||
AF061444.1 | - | 18 | 2567 | |||
AF062006.1 | - | 18 | 2615 | |||
AK075399.1 | - | 20 | 2366 | |||
AK315133.1 | - | 18 | 2855 | |||
BC096324.3 | - | 18 | 2613 | 40008253 | ||
BC096325.1 | - | 17 | 2541 | 40008255 | ||
BC096326.3 | - | 17 | 2541 | 40008257 | ||
BC099650.1 | - | 18 | 2613 | 40008256 | ||
FN820440.1 | - | 17 | 2399 |