Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014073.1 | NP_001014095.1 | 4 | 510 | ||
GenBank mRNA | BC089217.1 | - | 4 | 510 | 7304253 |