Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108009.1 | NP_001101479.1 | 6 | 627 | ||
XM_006239757.2 | XP_006239819.1 | 6 | 627 | |||
XM_006239758.2 | XP_006239820.1 | 7 | 1064 | |||
XM_006239759.2 | XP_006239821.1 | 6 | 956 | |||
XM_006239760.2 | XP_006239822.1 | 6 | 931 | |||
XM_006239761.2 | XP_006239823.1 | 4 | 452 | |||
XM_006239762.2 | XP_006239824.1 | 5 | 823 | |||
XM_008764485.1 | XP_008762707.1 | 2 | 1122 |