Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319210.1 | NP_001306139.1 | 4, 5 | 798 | ||
NM_001319211.1 | NP_001306140.1 | 4, 5 | 430 | |||
NM_001319212.1 | NP_001306141.1 | 4, 5 | 798 | |||
NM_003672.3 | NP_003663.2 | 4, 5 | 798 | |||
NM_033312.2 | NP_201569.1 | 4, 5 | 798 | |||
NM_033313.2 | NP_201570.1 | 4, 5 | 798 | |||
XM_005271294.3 | XP_005271351.1 | 4, 5 | 338 | |||
XM_005271296.3 | XP_005271353.1 | 4, 5 | 335 | |||
XM_011542340.2 | XP_011540642.1 | 4, 5 | 337 | |||
XM_011542341.2 | XP_011540643.1 | 3, 4 | 278 | |||
XM_011542346.2 | XP_011540648.1 | 4, 5 | 346 | |||
XM_017002646.1 | XP_016858135.1 | 4, 5 | 338 | |||
XM_017002647.1 | XP_016858136.1 | 4, 5 | 879 | |||
GenBank mRNA | AB209857.1 | - | 1, 2 | 227 | ||
AF000367.1 | - | 4, 5 | 665 | |||
AF064102.1 | - | 4, 5 | 773 | |||
AF064103.1 | - | 4, 5 | 763 | |||
AF122013.1 | - | 4, 5 | 310 | |||
AK302310.1 | - | 4, 5 | 419 | |||
AK310292.1 | - | 3, 4 | 316 | |||
BC038979.1 | - | 4, 5 | 372 | |||
BC093916.1 | - | 4, 5 | 405 | 7939761 | ||
BC093918.1 | - | 4, 5 | 405 | 7939763 | ||
DB029799.1 | - | 4, 5 | 443 | |||
DQ530256.1 | - | 4, 5 | 316 |