Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_233975.8 | XP_233975.5 | 3 | 306 | ||
GenBank mRNA | FQ224595.1 | - | 3 | 275 |