Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014076.1 | NP_001014098.1 | 16 | 1339 | ||
GenBank mRNA | BC081954.1 | - | 16 | 1339 | 7122941 |