Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003673.3 | NP_003664.1 | 2 | 615 | ||
GenBank mRNA | AJ000491.1 | - | 2 | 611 | ||
AK096328.1 | - | 2 | 1774 | |||
BC012628.1 | - | 2 | 616 | 4251080 | ||
BC013330.1 | - | 2 | 616 | 4273711 |