Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240009.2 | XP_006240071.1 | 9 | 1495 | ||
XM_006240013.2 | XP_006240075.1 | 8 | 1767 | |||
XM_006240014.2 | XP_006240076.1 | 8 | 1975 | |||
XM_006240015.2 | XP_006240077.1 | 9 | 1495 | |||
XM_006240016.2 | XP_006240078.1 | 9 | 1495 | |||
XM_006240017.2 | XP_006240079.1 | 7 | 1148 | |||
XM_006240018.2 | XP_006240080.1 | 6 | 1032 | |||
XM_008764632.1 | XP_008762854.1 | 9 | 1495 | |||
XM_008764633.1 | XP_008762855.1 | 8 | 2850 | |||
XM_008764634.1 | XP_008762856.1 | 9 | 1495 | |||
XM_008764635.1 | XP_008762857.1 | 6 | 1671 |