Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108021.1 | NP_001101491.1 | 3 | 967 | ||
XM_006257022.2 | XP_006257084.1 | 6 | 1242 | |||
XM_006257023.2 | XP_006257085.1 | 3 | 1367 | |||
XM_006257024.2 | XP_006257086.1 | 4 | 1418 | |||
XM_006257027.2 | XP_006257089.1 | 3 | 2296 | |||
XM_008773257.1 | XP_008771479.1 | 2 | 3087 | |||
XM_008773259.1 | XP_008771481.1 | 5 | 1462 | |||
XM_008773260.1 | XP_008771482.1 | 2 | 3087 |
Gene Symbol: LOC100911168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257037.1 | XP_006257099.1 | 2 | 198 |