Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750171.2 | XP_003750219.1 | 4 | 630 | ||
XM_006240140.2 | XP_006240202.1 | 4 | 1138 | |||
XM_006240141.2 | XP_006240203.1 | 5 | 1035 | |||
XM_006240142.2 | XP_006240204.1 | 4 | 700 | |||
XM_006240143.2 | XP_006240205.1 | 4 | 348 | |||
XM_006240144.2 | XP_006240206.1 | 3 | 284 | |||
GenBank mRNA | BC166472.1 | - | 3 | 1260 |