Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240169.2 | XP_006240231.1 | 11 | 967 | ||
XM_006240170.2 | XP_006240232.1 | 10 | 877 | |||
XM_008764716.1 | XP_008762938.1 | 11 | 967 | |||
XM_008764717.1 | XP_008762939.1 | 11 | 875 | |||
GenBank mRNA | BC159403.1 | - | 12 | 1156 | 7457550 |