Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025121.1 | NP_001020292.1 | 5 | 741 | ||
XM_006240171.2 | XP_006240233.1 | 6 | 1859 | |||
XM_006240172.2 | XP_006240234.1 | 6 | 1006 | |||
XM_006240173.2 | XP_006240235.1 | 3 | 1264 | |||
GenBank mRNA | BC081896.1 | - | 5 | 741 | 7110321 |