Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002877.1 | NP_001002877.1 | 8 | 828 | ||
NM_001002878.1 | NP_001002878.1 | 8 | 959 | |||
NM_001002879.1 | NP_001002879.1 | 7 | 843 | |||
NM_003678.4 | NP_003669.4 | 7 | 712 | |||
XM_005261797.1 | XP_005261854.1 | 7 | 725 | |||
XM_005261798.1 | XP_005261855.1 | 8 | 841 | |||
XM_005261799.1 | XP_005261856.1 | 7 | 868 | |||
XM_005261801.3 | XP_005261858.1 | 7 | 666 | |||
XM_006724347.1 | XP_006724410.1 | 8 | 955 | |||
XM_017029027.1 | XP_016884516.1 | 8 | 1327 | |||
GenBank mRNA | AB023200.1 | - | 7 | 679 | ||
AK025385.1 | - | 7 | 662 | |||
AK098709.1 | - | 4 | 329 | |||
AK122673.1 | - | 8 | 1335 | |||
AK225169.1 | - | 6 | 621 | |||
BC003615.2 | - | 7 | 652 | 2988049 | ||
BX381026.2 | - | 8 | 794 | |||
BX404329.2 | - | 7 | 716 | |||
CR456542.1 | - | 6 | 646 |