Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002877.1 | NP_001002877.1 | 4 | 411 | ||
NM_001002878.1 | NP_001002878.1 | 4 | 542 | |||
NM_001002879.1 | NP_001002879.1 | 3 | 426 | |||
NM_003678.4 | NP_003669.4 | 3 | 295 | |||
XM_005261797.1 | XP_005261854.1 | 3 | 308 | |||
XM_005261798.1 | XP_005261855.1 | 4 | 424 | |||
XM_005261799.1 | XP_005261856.1 | 3 | 451 | |||
XM_005261801.3 | XP_005261858.1 | 3 | 249 | |||
XM_006724347.1 | XP_006724410.1 | 4 | 538 | |||
XM_017029027.1 | XP_016884516.1 | 4 | 910 | |||
GenBank mRNA | AB023200.1 | - | 3 | 262 | ||
AK025385.1 | - | 3 | 245 | |||
AK122673.1 | - | 4 | 918 | |||
AK225169.1 | - | 2 | 204 | |||
AK225752.1 | - | 3 | 278 | |||
BC003615.2 | - | 3 | 235 | 2988049 | ||
BU187531.1 | - | 3 | 431 | |||
BX381026.2 | - | 4 | 376 | |||
BX404329.2 | - | 3 | 299 | |||
CN387648.1 | - | 4 | 547 | |||
CR456542.1 | - | 2 | 229 |