Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022197.2 | NP_071533.1 | 3, 4 | 660 | ||
GenBank mRNA | FQ225996.1 | - | 2, 3 | 313 | ||
FQ228884.1 | - | 2, 3 | 313 | |||
X06769.1 | - | 642 |