Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022197.2 | NP_071533.1 | 3 | 572 | ||
GenBank mRNA | FQ228884.1 | - | 2 | 225 | ||
X06769.1 | - | 554 |