Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem63c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108045.1 | NP_001101515.1 | 20 | 2200 | ||
XM_006240382.2 | XP_006240444.1 | 19 | 2188 | |||
XM_006240383.2 | XP_006240445.1 | 21 | 2342 | |||
XM_006240384.2 | XP_006240446.1 | 20 | 1973 | |||
XM_006240385.2 | XP_006240447.1 | 20 | 1977 | |||
XM_006240386.2 | XP_006240448.1 | 20 | 2023 | |||
XM_006240387.2 | XP_006240449.1 | 20 | 2394 | |||
XM_006240389.2 | XP_006240451.1 | 18 | 2040 | |||
XM_006240390.1 | XP_006240452.1 | 15 | 1378 | |||
XM_008764798.1 | XP_008763020.1 | 20 | 2251 | |||
GenBank mRNA | FQ212398.1 | - | 19 | 1891 |