Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108048.1 | NP_001101518.1 | 6, 7 | 857 | ||
XM_006240437.2 | XP_006240499.1 | 8 | 1040 | |||
XM_006240438.2 | XP_006240500.1 | 7 | 968 |