Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108049.1 | NP_001101519.1 | 1 | 1167 |
Gene Symbol: LOC102553138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240453.2 | XP_006240515.1 | 1 | 1336 |
Set Membership: |
G-protein coupled receptor |