Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003683.5 | NP_003674.1 | 8 | 746 | ||
XM_017028485.1 | XP_016883974.1 | 8 | 737 | |||
XM_017028486.1 | XP_016883975.1 | 8 | 728 | |||
GenBank mRNA | AK223185.1 | - | 8 | 707 | ||
AK223215.1 | - | 8 | 703 | |||
AK303000.1 | - | 8 | 706 | |||
AK313116.1 | - | 8 | 703 | |||
AL137757.1 | - | 5 | 767 | |||
AY033999.1 | - | 3 | 292 | |||
BC000380.2 | - | 8 | 693 | 2820455 | ||
BC014787.1 | - | 8 | 668 | |||
BI223794.1 | - | 3 | 120 | |||
U79775.1 | - | 8 | 691 |