Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papola | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108056.2 | NP_001101526.1 | 8 | 833 | ||
XM_006240521.2 | XP_006240583.1 | 8 | 843 | |||
XM_008764807.1 | XP_008763029.1 | 8 | 843 | |||
XM_008764808.1 | XP_008763030.1 | 8 | 843 | |||
XM_008764809.1 | XP_008763031.1 | 8 | 843 | |||
XM_008764810.1 | XP_008763032.1 | 8 | 842 | |||
GenBank mRNA | BC167010.1 | - | 8 | 787 | ||
FQ229237.1 | - | 8 | 786 |