Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100894.1 | NP_001094364.1 | 3 | 1055 | ||
XM_006240583.2 | XP_006240645.1 | 3 | 991 | |||
XM_006240585.2 | XP_006240647.1 | 2 | 808 | |||
XM_006240586.2 | XP_006240648.1 | 2 | 808 | |||
XM_008764933.1 | XP_008763155.1 | 4 | 1185 | |||
XM_008764934.1 | XP_008763156.1 | 4 | 1134 |