Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240617.2 | XP_006240679.1 | 6 | 1393 | ||
XM_006240618.2 | XP_006240680.1 | 6 | 1097 | |||
XM_006240620.2 | XP_006240682.1 | 5 | 809 | |||
XM_008764970.1 | XP_008763192.1 | 7 | 1160 | |||
XM_008764971.1 | XP_008763193.1 | 4 | 836 | |||
XM_008764972.1 | XP_008763194.1 | 5 | 779 |