Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012112.1 | NP_001012112.1 | 3 | 480 | ||
XM_008764936.1 | XP_008763158.1 | 2 | 1028 | |||
XM_008764937.1 | XP_008763159.1 | 3 | 355 | |||
GenBank mRNA | BC061827.1 | - | 1 | 263 | 5599008 | |
BC079192.1 | - | 3 | 480 | 7131342 |