Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rd3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134560.1 | NP_001128032.1 | 2 | 650 | ||
GenBank mRNA | BC158708.1 | - | 2 | 635 |