Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033702.1 | NP_001028874.1 | 8 | 858 | ||
XM_006240899.1 | XP_006240961.1 | 8 | 808 | |||
GenBank mRNA | BC099810.1 | - | 8 | 858 | 5621533 |