Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170557.1 | NP_001164028.1 | 14 | 1657 | ||
XM_006240921.2 | XP_006240983.1 | 14 | 1668 | |||
XM_006240922.2 | XP_006240984.1 | 14 | 1611 | |||
XM_006240923.2 | XP_006240985.1 | 13 | 1551 | |||
XM_008765098.1 | XP_008763320.1 | 14 | 1668 | |||
XM_008765099.1 | XP_008763321.1 | 15 | 1640 | |||
XM_008765100.1 | XP_008763322.1 | 14 | 1798 |