Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170557.1 | NP_001164028.1 | 5 | 579 | ||
XM_006240921.2 | XP_006240983.1 | 5 | 533 | |||
XM_006240922.2 | XP_006240984.1 | 5 | 533 | |||
XM_006240923.2 | XP_006240985.1 | 4 | 416 | |||
XM_006240925.2 | XP_006240987.1 | 5 | 533 | |||
XM_006240926.2 | XP_006240988.1 | 5 | 533 | |||
XM_008765098.1 | XP_008763320.1 | 5 | 533 | |||
XM_008765099.1 | XP_008763321.1 | 6 | 505 | |||
XM_008765100.1 | XP_008763322.1 | 5 | 663 |