Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012114.2 | NP_001012114.2 | 6 | 2478 | ||
GenBank mRNA | BC083596.1 | - | 2 | 473 |