Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108072.2 | NP_001101542.1 | 4 | 301 | ||
XM_006240928.1 | XP_006240990.1 | 4 | 328 | |||
XM_006240929.1 | XP_006240991.1 | 4 | 328 | |||
XM_006240930.1 | XP_006240992.1 | 4 | 328 | |||
GenBank mRNA | BC088451.1 | - | 2 | 1345 | ||
BC168179.1 | - | 4 | 301 | 9092463 | ||
FQ230285.1 | - | 4 | 570 |