Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003693.3 | NP_003684.2 | 11 | 3236 | ||
NM_145350.2 | NP_663325.1 | 11 | 3199 | |||
NR_028075.2 | 11 | 3166 | ||||
NR_102409.1 | 11 | 3273 | ||||
XM_017025256.1 | XP_016880745.1 | 7 | 2137 | |||
GenBank mRNA | AB052947.1 | - | 11 | 3099 | ||
BC039735.1 | - | 11 | 3202 | 6041360 | ||
D63483.1 | - | 11 | 3145 | |||
D86864.1 | - | 11 | 3169 |