Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291978.1 | NP_001278907.1 | 11, 12 | 1819 | ||
NM_001291979.1 | NP_001278908.1 | 11, 12 | 1819 | |||
NM_003703.2 | NP_003694.1 | 11, 12 | 1819 | |||
GenBank mRNA | AB000467.1 | - | 9, 10 | 1215 | ||
AF040965.1 | - | 11, 12 | 1681 | |||
AK024755.1 | - | 1, 2 | 128 | |||
AK025692.1 | - | 5, 6 | 684 | |||
AK292939.1 | - | 11, 12 | 1819 | |||
BC009760.2 | - | 7, 8 | 1013 | |||
BC026035.1 | - | 11, 12 | 1646 | 4878317 | ||
BC053635.1 | - | 11, 12 | 1698 | 5114280 |