Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241741.2 | XP_006241803.1 | 5 | 2122 | ||
XM_008765604.1 | XP_008763826.1 | 4 | 2406 | |||
XM_008765605.1 | XP_008763827.1 | 5 | 4377 |