Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUVBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319084.1 | NP_001306013.1 | 9 | 1240 | ||
NM_001319086.1 | NP_001306015.1 | 9 | 1648 | |||
NM_003707.2 | NP_003698.1 | 9 | 1181 | |||
XM_005247841.3 | XP_005247898.1 | 9 | 1102 | |||
XM_011513248.1 | XP_011511550.1 | 10 | 1335 | |||
XM_011513249.2 | XP_011511551.1 | 9 | 1235 | |||
XM_017007356.1 | XP_016862845.1 | 9 | 1235 | |||
XM_017007357.1 | XP_016862846.1 | 9 | 1105 | |||
GenBank mRNA | AB012122.1 | - | 9 | 1130 | ||
AB451224.1 | - | 9 | 1077 | |||
AF070735.1 | - | 9 | 1153 | |||
AF099084.1 | - | 9 | 1141 | |||
AJ010058.1 | - | 9 | 1153 | |||
AK092163.1 | - | 9 | 1648 | |||
AK222563.1 | - | 9 | 1162 | |||
AK222575.1 | - | 9 | 1153 | |||
AK312290.1 | - | 9 | 1177 | |||
BC002993.1 | - | 9 | 1148 | 2823568 | ||
BC012886.1 | - | 9 | 1110 | 3876400 | ||
BT007057.1 | - | 9 | 1077 | |||
DQ469310.1 | - | 9 | 1077 | |||
Y18418.1 | - | 9 | 1173 |