Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241927.2 | XP_006241989.1 | 5 | 638 | ||
XM_006241928.2 | XP_006241990.1 | 5 | 635 | |||
XM_008765571.1 | XP_008763793.1 | 5 | 635 |
Gene Symbol: Znf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750369.3 | XP_003750417.2 | 5 | 638 | ||
XM_006241919.2 | XP_006241981.1 | 5 | 635 | |||
XM_008765633.1 | XP_008763855.1 | 5 | 635 |