Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750385.3 | XP_003750433.1 | 13 | 8207 | ||
XM_006242130.2 | XP_006242192.1 | 14 | 3264 | |||
XM_008765806.1 | XP_008764028.1 | 13 | 3877 | |||
XM_235497.8 | XP_235497.6 | 14 | 2938 |