Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750385.3 | XP_003750433.1 | 4 | 5905 | ||
XM_006242130.2 | XP_006242192.1 | 5 | 962 | |||
XM_008765806.1 | XP_008764028.1 | 4 | 1575 | |||
XM_235497.8 | XP_235497.6 | 5 | 636 | |||
GenBank mRNA | AB588919.1 | - | 5 | 523 | ||
AB588920.1 | - | 4 | 378 | |||
AB588921.1 | - | 5 | 505 |