Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003713.4 | NP_003704.3 | 3 | 1096 | ||
GenBank mRNA | AB000889.1 | - | 3 | 534 | ||
AF017786.1 | - | 4 | 902 | |||
AF043329.1 | - | 3 | 821 | |||
AF480883.1 | - | 4 | 908 | |||
AK312439.1 | - | 3 | 1054 | |||
BC009196.2 | - | 3 | 899 | 3960223 | ||
BT019589.1 | - | 3 | 528 | |||
U79294.1 | - | 4 | 902 |