Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108112.1 | NP_001101582.1 | 4 | 538 | ||
XM_006257400.2 | XP_006257462.1 | 4 | 485 | |||
GenBank mRNA | BC166473.1 | - | 4 | 438 | 9090730 |