Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290049.1 | NP_001276978.1 | 13, 14 | 1550 | ||
NM_003715.3 | NP_003706.2 | 12, 13 | 1538 | |||
XM_006714396.3 | XP_006714459.1 | 12, 13 | 1533 | |||
XM_006714397.3 | XP_006714460.1 | 13, 14 | 1550 | |||
GenBank mRNA | AK002093.1 | - | 7, 8 | 758 | ||
AK314289.1 | - | 12, 13 | 1428 | |||
AL832010.2 | - | 13, 14 | 1473 | |||
AL833753.1 | - | 2, 3 | 249 | |||
BC006398.2 | - | 12, 13 | 1463 | |||
BC032654.1 | - | 12, 13 | 1393 | 5533256 | ||
D86326.1 | - | 12, 13 | 1415 |