Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290049.1 | NP_001276978.1 | 10 | 1075 | ||
NM_003715.3 | NP_003706.2 | 9 | 1063 | |||
XM_006714396.3 | XP_006714459.1 | 9 | 1058 | |||
XM_006714397.3 | XP_006714460.1 | 10 | 1075 | |||
GenBank mRNA | AK002093.1 | - | 4 | 283 | ||
AK314289.1 | - | 9 | 953 | |||
AL832010.2 | - | 10 | 998 | |||
BC006398.2 | - | 9 | 988 | |||
BC032654.1 | - | 9 | 918 | 5533256 | ||
D86326.1 | - | 9 | 940 |