Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079707.1 | NP_001073175.1 | 3 | 376 | ||
XM_006242403.2 | XP_006242465.1 | 4 | 348 | |||
XM_008765750.1 | XP_008763972.1 | 4 | 338 | |||
XM_008765751.1 | XP_008763973.1 | 4 | 340 | |||
XM_008765752.1 | XP_008763974.1 | 4 | 409 | |||
GenBank mRNA | BC128778.1 | - | 3 | 376 | 7134533 |